Assay Details
Target Gene Details
Entrez Gene ID: | 790910 |
Gene Name: | predicted gene 11201 |
Gene Aliases: |
ENSMUSG00000072637, OTTMUSG00000000169 |
Location: |
Chr.11:79023597-79035005 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11201 | NR_045873.1 | |||
| AK154065.1 | BAE32351.1 |
Target Gene Details
Entrez Gene ID: | 16706 |
Gene Name: | kinase suppressor of ras 1 |
Gene Aliases: |
AW492498, B-KSR1, D11Bhm183e, D11Bhm184e, Ksr, RSU2, hb |
Location: |
Chr.11:79013440-79146554 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ksr1 | NM_013571.2 | NP_038599.1 | ||
| AK036566.1 | ||||
| AK137532.1 | BAE23398.1 | |||
| U43585.1 | AAC52382.1 |
More Information
Set Membership: |
|

Genomic Map