Assay Details
Target Gene Details
Entrez Gene ID: | 632687 |
Gene Name: | membrane-associated ring finger (C3HC4) 10 |
Gene Aliases: |
4933417C16Rik, AV039223, OTTMUSG00000002847, Rnf190 |
Location: |
Chr.11:105360798-105456863 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March10 | NM_172568.2 | NP_766156.2 | ||
| AK077168.1 | BAC36658.1 | |||
| BC117499.1 |
More Information
Set Membership: |
|

Genomic Map