Assay Details
Target Gene Details
Entrez Gene ID: | 21428 |
Gene Name: | MAX-like protein X |
Gene Aliases: |
Tcfl4, bHLHd13 |
Location: |
Chr.11:101086709-101092456 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlx | NM_001159384.1 | 6 | 519 | NP_001152856.1 |
| NM_001159385.1 | 7 | 609 | NP_001152857.1 | |
| NM_011550.3 | 7 | 771 | NP_035680.3 | |
| AF213670.1 | 6 | 483 | AAG40149.1 | |
| AF213671.1 | 7 | 573 | AAG40150.1 | |
| AF213672.1 | 7 | 735 | AAG40151.1 | |
| AK011478.1 | 6 | 499 | ||
| AK013973.1 | 1 | 44 | ||
| AK142605.1 | 7 | 575 | BAE25128.1 | |
| BC031380.1 | 7 | 751 | AAH31380.1 | |
| U43548.1 | 7 | 731 | AAB51367.1 | |
| U43607.1 | 7 | 742 | AAB51368.1 |
More Information
Set Membership: |
|

Genomic Map