Assay Details
Target Gene Details
Entrez Gene ID: | 216767 |
Gene Name: | mitochondrial ribosomal protein L22 |
Gene Aliases: |
E030011D16Rik, HSPC158, MRP-L25, Rpml25 |
Location: |
Chr.11:58171604-58179580 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl22 | NM_175001.3 | NP_778166.2 | ||
| AK077615.1 | BAC36901.1 | |||
| AK086913.1 | BAC39761.1 | |||
| AK164417.1 | BAE37779.1 | |||
| BC061191.1 | AAH61191.1 | |||
| BY790081.1 |
More Information
Set Membership: |
|

Genomic Map