Assay Details
Target Gene Details
Entrez Gene ID: | 268465 |
Gene Name: | essential meiotic endonuclease 1 homolog 1 (S. pombe) |
Gene Aliases: |
6820428D13 |
Location: |
Chr.11:94644998-94653785 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eme1 | NM_177752.4 | NP_808420.1 | ||
| AK078516.1 | BAC37318.1 | |||
| BC089459.1 | AAH89459.1 |
Target Gene Details
Entrez Gene ID: | 94064 |
Gene Name: | mitochondrial ribosomal protein L27 |
Gene Aliases: |
A630055F16Rik, D11Moh47, D18Ertd643e |
Location: |
Chr.11:94653770-94660087 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl27 | NM_053161.2 | NP_444391.1 | ||
| AB049648.1 | BAB40853.1 | |||
| AK080324.1 | ||||
| AK080329.1 | ||||
| AK143575.1 | ||||
| BC030074.1 | AAH30074.1 |
More Information
Set Membership: |
|

Genomic Map