Assay Details
Target Gene Details
Entrez Gene ID: | 217143 |
Gene Name: | G protein-coupled receptor 179 |
Gene Aliases: |
5330439C02Rik, Gpr158l1 |
Location: |
Chr.11:97332109-97352394 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr179 | NM_001081220.1 | 6 | 1408 | NP_001074689.1 |
More Information
Set Membership: |
|

Genomic Map