Assay Details
Target Gene Details
Entrez Gene ID: | 77605 |
Gene Name: | H2A histone family, member V |
Gene Aliases: |
C530002L11Rik, H2a.z-2, H2av |
Location: |
Chr.11:6427226-6444443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2afv | NM_029938.1 | NP_084214.1 | ||
| AK021267.1 | BAB32354.1 | |||
| AK155105.1 | BAE33050.1 | |||
| AK168110.1 | BAE40081.1 | |||
| BC028539.1 | AAH28539.1 | |||
| BC147293.1 | ||||
| BC147294.1 | ||||
| BC147741.1 | ||||
| BC147751.1 | ||||
| CJ069840.1 |
More Information
Set Membership: |
|

Genomic Map