Assay Details
Target Gene Details
Entrez Gene ID: | 360013 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MAJN, MyoPDZ, MysPDZ |
Location: |
Chr.11:77763251-77865988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo18a | NM_001291212.1 | NP_001278141.1 | ||
| NM_001291213.1 | NP_001278142.1 | |||
| NM_001291214.1 | NP_001278143.1 | |||
| NM_001291215.1 | NP_001278144.1 | |||
| NM_011586.3 | NP_035716.1 | |||
| AB026497.1 | BAA93660.1 | |||
| AK137574.1 | BAE23413.1 | |||
| AK147584.1 | BAE28009.1 | |||
| AK171342.1 | BAE42402.1 | |||
| BC138366.1 |

Genomic Map