Assay Details
Target Gene Details
Entrez Gene ID: | 16796 |
Gene Name: | LIM and SH3 protein 1 |
Gene Aliases: |
AA408629, Def-4, SH3P6, Tg(Col1a1-lacZ)1Ngma |
Location: |
Chr.11:97799005-97838765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lasp1 | NM_010688.4 | NP_034818.1 | ||
| AK034404.1 | BAC28696.1 | |||
| AK049199.1 | BAC33604.1 | |||
| AK078445.1 | BAC37278.1 | |||
| AK148858.1 | BAE28681.1 | |||
| AK154656.1 | BAE32746.1 | |||
| AK168521.1 | BAE40400.1 | |||
| AK169381.1 | BAE41128.1 | |||
| BC010840.1 | AAH10840.1 | |||
| CT010288.1 | CAJ18496.1 | |||
| U58882.1 | AAC52639.1 | |||
| X96973.1 | CAA65659.1 |

Genomic Map