Assay Details
Target Gene Details
Entrez Gene ID: | 268395 |
Gene Name: | N-methylpurine-DNA glycosylase |
Gene Aliases: |
9830006D05, AI326268, APNG, Aag, Mid1 |
Location: |
Chr.11:32226425-32232702 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpg | NM_010822.3 | NP_034952.2 | ||
| AK018694.1 | ||||
| AK076201.1 | ||||
| AK088878.1 | ||||
| AK134900.1 | ||||
| AK155766.1 | ||||
| AK156067.1 | BAE33569.1 | |||
| BC014754.1 | AAH14754.1 | |||
| U10420.1 | AAA19487.1 |
More Information
Set Membership: |
|

Genomic Map