Assay Details
Target Gene Details
Entrez Gene ID: | 16878 |
Gene Name: | leukemia inhibitory factor |
Gene Aliases: |
- |
Location: |
Chr.11:4257568-4272514 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lif | NM_008501.2 | 2 | 179 | NP_032527.1 |
| AF065917.1 | AAC17912.1 | |||
| AF065918.1 | AAC17913.1 | |||
| AK155959.1 | BAE33523.1 | |||
| BY207460.1 | 2 | 180 | ||
| BY209921.1 | ||||
| X12810.1 | CAA31299.1 |
More Information
Set Membership: |
|

Genomic Map