Assay Details
Target Gene Details
Entrez Gene ID: | 17913 |
Gene Name: | myosin IC |
Gene Aliases: |
C80397, MMIb, MYO1E, NMI, mm1beta, myr2 |
Location: |
Chr.11:75650493-75674643 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo1c | NM_001080774.1 | 2 | 278 | NP_001074243.1 |
| NM_001080775.1 | 2 | 271 | NP_001074244.1 | |
| NM_008659.3 | 2 | 241 | NP_032685.1 | |
| AK004743.1 | 2 | 170 | BAB23524.1 | |
| AK154294.1 | 2 | 137 | BAE32495.1 | |
| AK155530.1 | 2 | 217 | BAE33311.1 | |
| AK165680.1 | 2 | 137 | ||
| AK166638.1 | 2 | 279 | ||
| AK171107.1 | 2 | 272 | BAE42253.1 | |
| AY007255.1 | 2 | 127 | AAG02570.1 | |
| BC021481.1 | 2 | 182 | AAH21481.1 | |
| BY310607.1 | 2 | 241 | ||
| CK020066.1 | 2 | 245 | ||
| U96723.1 | 1 | 65 | AAC53264.1 | |
| X99638.1 | 2 | 200 | CAA67956.1 |
More Information
Set Membership: |
|

Genomic Map