Assay Details
Target Gene Details
Entrez Gene ID: | 16882 |
Gene Name: | ligase III, DNA, ATP-dependent |
Gene Aliases: |
D11Wsu78e |
Location: |
Chr.11:82781109-82804274 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig3 | NM_001291245.1 | 4 | 961 | NP_001278174.1 |
| NM_001291246.1 | 4 | 958 | NP_001278175.1 | |
| NM_001291247.1 | 4 | 962 | NP_001278176.1 | |
| NM_010716.3 | 4 | 965 | NP_034846.2 | |
| AK010719.1 | 4 | 876 | ||
| AK031540.1 | 4 | 903 | ||
| AK143531.1 | 4 | 831 | BAE25421.1 | |
| AK150644.1 | 4 | 870 | BAE29732.1 | |
| AK156018.1 | 4 | 880 | BAE33548.1 | |
| AK167363.1 | 4 | 873 | BAE39460.1 | |
| BC049240.1 | 4 | 868 | AAH49240.1 | |
| BC083500.1 | 4 | 864 | AAH83500.1 | |
| U66057.1 | 4 | 890 | AAC53003.1 | |
| U66058.1 | 4 | 890 | AAC53004.1 |
More Information
Set Membership: |
|

Genomic Map