Assay Details
Target Gene Details
Entrez Gene ID: | 104601 |
Gene Name: | MYCBP associated protein |
Gene Aliases: |
4932408B01Rik, AMAP-1, AW125474 |
Location: |
Chr.11:94501347-94532388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mycbpap | NM_170671.2 | NP_733771.2 | ||
| AK029929.1 | BAC26684.1 | |||
| AK166780.1 | ||||
| BC027327.1 | AAH27327.1 | |||
| BC050808.1 | AAH50808.1 |

Genomic Map