Assay Details
Target Gene Details
Entrez Gene ID: | 104601 |
Gene Name: | MYCBP associated protein |
Gene Aliases: |
4932408B01Rik, AMAP-1, AW125474 |
Location: |
Chr.11:94501347-94532388 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mycbpap | NM_170671.2 | 19 | 2796 | NP_733771.2 |
| AK029929.1 | 18 | 2972 | BAC26684.1 | |
| AK166780.1 | 12 | 1944 | ||
| BC027327.1 | 15 | 2157 | AAH27327.1 | |
| BC050808.1 | 8 | 1319 | AAH50808.1 | |
| BE979961.1 | 1 | 148 |

Genomic Map