Assay Details
Target Gene Details
Entrez Gene ID: | 15239 |
Gene Name: | HGF-regulated tyrosine kinase substrate |
Gene Aliases: |
Hgr, Hrs, ZFYVE8 |
Location: |
Chr.11:120467635-120483984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgs | NM_001159328.1 | NP_001152800.1 | ||
| NM_008244.3 | NP_032270.3 | |||
| AK046299.1 | BAC32676.1 | |||
| AK145031.1 | BAE26195.1 | |||
| AK166441.1 | BAE38778.1 | |||
| AK180016.1 | ||||
| AK206792.1 | ||||
| AK210546.1 | ||||
| AK215004.1 | ||||
| AK216932.1 | ||||
| AK217735.1 | ||||
| BC003239.1 | AAH03239.1 | |||
| D50050.1 | BAA08768.1 |
More Information
Set Membership: |
|

Genomic Map