Assay Details
Target Gene Details
Entrez Gene ID: | 216850 |
Gene Name: | KDM1 lysine (K)-specific demethylase 6B |
Gene Aliases: |
1700064E03Rik, BC038313, Jmjd3, mKIAA0346 |
Location: |
Chr.11:69398508-69418148 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm6b | NM_001017426.1 | 20 | 4942 | NP_001017426.1 |
| AK129121.1 | 10 | 1734 | BAC97931.1 | |
| AK136019.1 | 10 | 1123 | BAE22775.1 | |
| BC029201.1 | AAH29201.1 | |||
| BC075632.1 | 21 | 5118 | AAH75632.1 |
More Information
Set Membership: |
|

Genomic Map