Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogdh | NM_001252282.1 | NP_001239211.1 | ||
| NM_001252283.1 | NP_001239212.1 | |||
| NM_001252287.1 | NP_001239216.1 | |||
| NM_001252288.1 | NP_001239217.1 | |||
| NM_010956.4 | NP_035086.2 | |||
| AK147289.1 | BAE27824.1 | |||
| AK150009.1 | BAE29234.1 | |||
| AK169286.1 | BAE41044.1 | |||
| AK220536.1 | BAD90530.1 | |||
| BC013670.1 | AAH13670.1 | |||
| BC049104.1 | AAH49104.1 | |||
| BC057354.1 | AAH57354.1 | |||
| BC061232.1 | AAH61232.1 |
More Information
Set Membership: |
|

Genomic Map