Assay Details
Target Gene Details
Entrez Gene ID: | 276846 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class S |
Gene Aliases: |
BC058979, Gm393, Gm689 |
Location: |
Chr.11:78328422-78342781 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigs | NM_201406.1 | 11 | 1380 | NP_958808.1 |
| AK033331.1 | 11 | 1354 | BAE20485.1 | |
| AK039604.1 | 11 | 1351 | BAE20546.1 | |
| AK043232.1 | 11 | 1354 | BAE20638.1 | |
| AK044292.1 | 4 | 2091 | ||
| AK045759.1 | 11 | 1350 | ||
| AK053771.1 | 11 | 1354 | BAE20693.1 | |
| AK142871.1 | 11 | 1351 | ||
| AK151858.1 | 11 | 1342 | BAE30747.1 | |
| AK152229.1 | 11 | 1342 | BAE31056.1 | |
| AK153300.1 | 11 | 1346 | BAE31882.1 | |
| AK166408.1 | 11 | 1357 | ||
| AK169633.1 | 10 | 1153 | ||
| AK180839.1 | 2 | 265 | ||
| AK216113.1 | 2 | 243 | ||
| BC058979.1 | 11 | 1380 | AAH58979.1 |

Genomic Map