Assay Details
Target Gene Details
Entrez Gene ID: | 18750 |
Gene Name: | protein kinase C, alpha |
Gene Aliases: |
AI875142, Pkca |
Location: |
Chr.11:107933387-108343888 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkca | NM_011101.3 | 9 | 1205 | NP_035231.2 |
| AK147084.1 | 9 | 1206 | BAE27664.1 | |
| BC096493.1 | 9 | 999 | AAH96493.1 | |
| M25811.1 | 9 | 1239 | AAA39934.1 | |
| X52684.1 | 10 | 1146 | CAA36907.1 | |
| X52685.1 | 10 | 1146 | CAA36908.1 |
More Information
Set Membership: |
|

Genomic Map