Assay Details
Target Gene Details
Entrez Gene ID: | 19127 |
Gene Name: | paired like homeodomain factor 1 |
Gene Aliases: |
Prop-1, df |
Location: |
Chr.11:50950806-50953632 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prop1 | NM_008936.1 | NP_032962.1 | ||
| U77946.1 | AAB38884.1 |
More Information
Set Membership: |
|

Genomic Map