Assay Details
Target Gene Details
Entrez Gene ID: | 50724 |
Gene Name: | SAP30-like |
Gene Aliases: |
2310079P12Rik, AF006998, L55 |
Location: |
Chr.11:57801637-57810615 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap30l | NM_001081168.1 | NP_001074637.1 | ||
| AK010234.1 | BAB26785.1 | |||
| AK203234.1 | ||||
| BC051686.1 | ||||
| BC094930.1 | AAH94930.1 | |||
| BC117029.1 | ||||
| BC117031.1 |
More Information
Set Membership: |
|

Genomic Map