Assay Details
Target Gene Details
Entrez Gene ID: | 66397 |
Gene Name: | SAR1 gene homolog B (S. cerevisiae) |
Gene Aliases: |
2310075M17Rik, 2900019I22Rik, CMRD, Sara1b, Sara2, Sarb |
Location: |
Chr.11:51763663-51791953 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sar1b | NM_025535.2 | NP_079811.1 | ||
| AK002327.1 | BAB22015.1 | |||
| AK010187.1 | BAB26755.1 | |||
| AK013559.1 | BAB28905.1 | |||
| AK150906.1 | BAE29948.1 | |||
| AK155429.1 | BAE33261.1 | |||
| BC082550.1 | AAH82550.1 | |||
| BC085178.1 | ||||
| BY068863.1 |

Genomic Map