Assay Details
Target Gene Details
Entrez Gene ID: | 215335 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 1 |
Gene Aliases: |
5830411H19Rik, AI839897, Pat1 |
Location: |
Chr.11:55204321-55236330 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc36a1 | NM_153139.4 | NP_694779.3 | ||
| AF453743.1 | AAM80480.1 | |||
| AY211262.1 | AAO37090.1 | |||
| BC138556.1 | ||||
| BC138558.1 |
More Information
Set Membership: |
|

Genomic Map