Assay Details
Target Gene Details
Entrez Gene ID: | 246049 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 2 |
Gene Aliases: |
A530067G19Rik, PAT2, Tramd1 |
Location: |
Chr.11:55158468-55185095 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc36a2 | NM_153170.3 | NP_694810.2 | ||
| AF453744.1 | AAM80481.1 | |||
| AF512429.1 | AAM44854.1 | |||
| AK041041.1 | BAC30795.1 | |||
| AK079053.1 | BAC37515.1 | |||
| AK086373.1 | BAC39656.1 | |||
| AK156409.1 | BAE33702.1 | |||
| AK160394.1 | BAE35764.1 | |||
| AY228366.1 | AAO74833.1 | |||
| BC044800.1 | AAH44800.1 |
More Information
Set Membership: |
|

Genomic Map