Assay Details
Target Gene Details
Entrez Gene ID: | 20415 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP |
Location: |
Chr.11:69610281-69618025 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shbg | NM_011367.2 | NP_035497.1 | ||
| AK010959.1 | ||||
| BC120785.1 | ||||
| BC120787.1 | ||||
| U85644.1 | AAB42088.1 |

Genomic Map