Assay Details
Target Gene Details
Entrez Gene ID: | 246049 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 2 |
Gene Aliases: |
A530067G19Rik, PAT2, Tramd1 |
Location: |
Chr.11:55158468-55185095 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc36a2 | NM_153170.3 | 8 | 1033 | NP_694810.2 |
| AF453744.1 | 8 | 986 | AAM80481.1 | |
| AF512429.1 | 8 | 1056 | AAM44854.1 | |
| AK041041.1 | 8 | 1025 | BAC30795.1 | |
| AK079053.1 | 8 | 998 | BAC37515.1 | |
| AK156409.1 | 8 | 1034 | BAE33702.1 | |
| AK160394.1 | 8 | 998 | BAE35764.1 | |
| BC044800.1 | 8 | 1033 | AAH44800.1 |
More Information
Set Membership: |
|

Genomic Map