Assay Details
Target Gene Details
Entrez Gene ID: | 215332 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 3 |
Gene Aliases: |
PAT3, TRAMD2, tramdorin2 |
Location: |
Chr.11:55124815-55151745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc36a3 | NM_172258.3 | NP_758462.2 | ||
| AF453745.1 | AAN76273.1 | |||
| AK132910.1 | ||||
| AY211261.1 | AAO37089.1 |
More Information
Set Membership: |
|

Genomic Map