Assay Details
Target Gene Details
Entrez Gene ID: | 18023 |
Gene Name: | nuclear factor, erythroid derived 2,-like 1 |
Gene Aliases: |
AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11 |
Location: |
Chr.11:96817413-96829968 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfe2l1 | NM_001130450.1 | NP_001123922.1 | ||
| NM_008686.3 | NP_032712.2 | |||
| AF071084.1 | AAC83235.1 | |||
| AK131183.1 | BAD21433.1 | |||
| AK152040.1 | BAE30899.1 | |||
| AK153529.1 | BAE32068.1 | |||
| AK154167.1 | BAE32418.1 | |||
| AK184212.1 | ||||
| AK201732.1 | ||||
| BC043063.1 | AAH43063.1 | |||
| BC047283.1 | AAH47283.1 | |||
| CJ152359.1 | ||||
| X78709.1 | CAA55362.1 |
More Information
Set Membership: |
|

Genomic Map