Assay Details
Target Gene Details
Entrez Gene ID: | 18023 |
Gene Name: | nuclear factor, erythroid derived 2,-like 1 |
Gene Aliases: |
AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11 |
Location: |
Chr.11:96817413-96829968 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfe2l1 | NM_008686.3 | NP_032712.2 | ||
| AK131183.1 | BAD21433.1 | |||
| AK152040.1 | BAE30899.1 | |||
| AK153529.1 | BAE32068.1 | |||
| BY301848.1 |
More Information
Set Membership: |
|

Genomic Map