Assay Details
Target Gene Details
Entrez Gene ID: | 18023 |
Gene Name: | nuclear factor, erythroid derived 2,-like 1 |
Gene Aliases: |
AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11 |
Location: |
Chr.11:96817413-96829968 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfe2l1 | NM_001130450.1 | 3 | 1230 | NP_001123922.1 |
| NM_001130451.1 | 2 | 164 | NP_001123923.1 | |
| NM_001130452.1 | 2 | 131 | NP_001123924.1 | |
| NM_001130453.1 | 2 | 181 | NP_001123925.1 | |
| NM_001130454.1 | 2 | 148 | NP_001123926.1 | |
| NM_008686.3 | 3 | 1288 | NP_032712.2 | |
| AF071084.1 | 2 | 634 | AAC83235.1 | |
| AK131183.1 | 3 | 1223 | BAD21433.1 | |
| AK148471.1 | 2 | 148 | BAE28572.1 | |
| AK152040.1 | 3 | 1269 | BAE30899.1 | |
| AK153529.1 | 3 | 1270 | BAE32068.1 | |
| AK154167.1 | 3 | 1199 | BAE32418.1 | |
| AK184212.1 | 2 | 298 | ||
| AK194563.1 | 1 | 110 | ||
| AK201732.1 | 2 | 289 | ||
| BC043063.1 | 2 | 870 | AAH43063.1 | |
| BC047283.1 | 2 | 630 | AAH47283.1 | |
| CF163519.1 | 2 | 176 | ||
| CJ144781.1 | 2 | 137 | ||
| CJ152359.1 | 2 | 152 | ||
| X78709.1 | 2 | 666 | CAA55362.1 |
More Information
Set Membership: |
|

Genomic Map