Assay Details
Target Gene Details
Entrez Gene ID: | 75273 |
Gene Name: | proline, glutamic acid and leucine rich protein 1 |
Gene Aliases: |
4930563C04Rik, MNAR |
Location: |
Chr.11:70392881-70410031 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pelp1 | NM_029231.4 | NP_083507.3 | ||
| AK005027.1 | BAB23754.1 | |||
| AK128966.1 | ||||
| BC016444.1 | AAH16444.1 | |||
| BC057987.1 | AAH57987.1 | |||
| BC090620.1 |

Genomic Map