Assay Details
Target Gene Details
Entrez Gene ID: | 72055 |
Gene Name: | solute carrier family 38, member 10 |
Gene Aliases: |
1810073N04Rik |
Location: |
Chr.11:120103951-120151351 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc38a10 | NM_001164798.1 | 8 | 1098 | NP_001158270.1 |
| NM_001164799.1 | 8 | 1098 | NP_001158271.1 | |
| NM_001164800.1 | 8 | 1098 | NP_001158272.1 | |
| NM_001164801.1 | 8 | 1098 | NP_001158273.1 | |
| NM_001164802.1 | 8 | 1098 | NP_001158274.1 | |
| NM_024249.5 | 8 | 1098 | NP_077211.4 | |
| AK007977.1 | 8 | 988 | BAB25384.1 | |
| AK032245.1 | 8 | 1022 | BAC27777.1 | |
| AK090050.1 | 8 | 1074 | BAC41066.1 | |
| AK159948.1 | 8 | 1060 | BAE35504.1 | |
| AK165672.1 | 8 | 1063 | ||
| AK180057.1 | ||||
| AK183661.1 | ||||
| BC082300.1 | 8 | 1069 | AAH82300.1 | |
| BC088811.1 | 8 | 1034 | AAH88811.1 |
More Information
Set Membership: |
|

Genomic Map