Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc43a2 | NM_001199284.1 | NP_001186213.1 | ||
| NM_173388.2 | NP_775564.1 | |||
| BY202700.1 |

Genomic Map