Assay Details
Target Gene Details
Entrez Gene ID: | 71966 |
Gene Name: | NFKB inhibitor interacting Ras-like protein 2 |
Gene Aliases: |
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2 |
Location: |
Chr.11:100622955-100627602 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkiras2 | NM_028024.2 | NP_082300.1 | ||
| AK010371.1 | BAB26889.1 | |||
| AK052645.1 | BAC35078.1 | |||
| AK083475.1 | BAC38929.1 | |||
| AK149696.1 | BAE29033.1 | |||
| AK169878.1 | BAE41430.1 | |||
| BC013469.1 | AAH13469.1 | |||
| BC024398.1 | AAH24398.1 |

Genomic Map