Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc43a2 | NM_001199283.1 | 8 | 1171 | NP_001186212.1 |
| NM_001199284.1 | 8 | 953 | NP_001186213.1 | |
| NM_173388.2 | 8 | 916 | NP_775564.1 | |
| AB120363.1 | 8 | 949 | BAD91090.1 | |
| AK143773.1 | 8 | 1014 | BAE25533.1 | |
| AK159144.1 | 8 | 1139 | BAE34852.1 | |
| AK171104.1 | 8 | 1172 | ||
| AK172685.1 | 8 | 943 | ||
| BC042513.1 | 8 | 912 | AAH42513.1 |

Genomic Map