Assay Details
Target Gene Details
Entrez Gene ID: | 71701 |
Gene Name: | polyribonucleotide nucleotidyltransferase 1 |
Gene Aliases: |
1200003F12Rik, Old35, PNPase, Pnptl1 |
Location: |
Chr.11:29130215-29162010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpt1 | NM_027869.1 | NP_082145.1 | ||
| AF465249.1 | AAO33354.1 | |||
| AJ507387.1 | CAD45436.1 | |||
| AK149419.1 | BAE28862.1 | |||
| AK152445.1 | ||||
| AK161833.1 | BAE36595.1 | |||
| AK165566.1 | BAE38260.1 | |||
| BC027228.1 | AAH27228.2 | |||
| BC055826.1 | AAH55826.1 |
More Information
Set Membership: |
|

Genomic Map