Assay Details
Target Gene Details
Entrez Gene ID: | 94045 |
Gene Name: | purinergic receptor P2X, ligand-gated ion channel, 5 |
Gene Aliases: |
P2X5 |
Location: |
Chr.11:73155277-73175327 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2rx5 | NM_033321.3 | NP_201578.2 | ||
| AF333331.1 | AAK49936.1 | |||
| AK134662.1 | BAE22231.1 | |||
| BC138189.1 | ||||
| BC145181.1 | ||||
| JX570808.1 |
More Information
Set Membership: |
|

Genomic Map