Assay Details
Target Gene Details
Entrez Gene ID: | 18738 |
Gene Name: | phosphatidylinositol transfer protein, alpha |
Gene Aliases: |
Pitpn, vb, vibrator |
Location: |
Chr.11:75588108-75628778 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpna | NM_008850.1 | NP_032876.1 | ||
| AK144618.1 | BAE25972.1 | |||
| AK149745.1 | BAE29058.1 | |||
| AK151496.1 | BAE30448.1 | |||
| AK152089.1 | BAE30939.1 | |||
| AK152874.1 | ||||
| AK153569.1 | BAE32103.1 | |||
| AK165659.1 | BAE38321.1 | |||
| AK167760.1 | BAE39793.1 | |||
| AK168187.1 | BAE40147.1 | |||
| AK168720.1 | BAE40562.1 | |||
| AK187087.1 | ||||
| BC056171.1 | AAH56171.1 | |||
| S72681.1 | AAC60690.1 | |||
| U96725.1 | AAC53266.1 |
More Information
Set Membership: |
|

Genomic Map