Assay Details
Target Gene Details
Entrez Gene ID: | 71701 |
Gene Name: | polyribonucleotide nucleotidyltransferase 1 |
Gene Aliases: |
1200003F12Rik, Old35, PNPase, Pnptl1 |
Location: |
Chr.11:29130215-29162010 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpt1 | NM_027869.1 | 4 | 363 | NP_082145.1 |
| AF465249.1 | 4 | 362 | AAO33354.1 | |
| AJ507387.1 | 4 | 363 | CAD45436.1 | |
| AK149419.1 | 4 | 364 | BAE28862.1 | |
| AK152445.1 | 4 | 340 | ||
| AK161833.1 | 4 | 364 | BAE36595.1 | |
| AK165566.1 | 4 | 366 | BAE38260.1 | |
| BC055826.1 | 4 | 396 | AAH55826.1 |
More Information
Set Membership: |
|

Genomic Map