Assay Details
Target Gene Details
Entrez Gene ID: | 71795 |
Gene Name: | phosphatidylinositol transfer protein, cytoplasmic 1 |
Gene Aliases: |
1110020B03Rik, 5830436L09Rik, AI662802, AI851387, C330017I21Rik, Dnr411, RDGBB, RDGBB1, rdgB-beta |
Location: |
Chr.11:107207892-107470892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitpnc1 | NM_145823.2 | NP_665822.1 | ||
| AB077281.1 | BAC02913.1 | |||
| AB077282.1 | BAC02914.1 | |||
| AK003844.1 | BAC25057.1 | |||
| AK021206.1 | ||||
| AK153625.1 | BAE32124.1 | |||
| AK171340.1 | BAE42401.1 | |||
| AK171918.1 | ||||
| AK172137.1 | ||||
| BC108351.1 | AAI08352.1 |
More Information
Set Membership: |
|

Genomic Map