Assay Details
Target Gene Details
Entrez Gene ID: | 18972 |
Gene Name: | polymerase (DNA directed), delta 2, regulatory subunit |
Gene Aliases: |
50kDa, p50, po1D2 |
Location: |
Chr.11:5872180-5878256 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pold2 | NM_008894.2 | 3 | 402 | NP_032920.2 |
| AK013901.1 | 3 | 369 | ||
| AK218431.1 | 2 | 117 | ||
| BC003342.1 | 3 | 368 | AAH03342.1 | |
| BG078511.2 | 3 | 355 | ||
| BY093766.1 | ||||
| Z72486.1 | 3 | 363 | CAA96567.1 |
More Information
Set Membership: |
|

Genomic Map