Assay Details
Target Gene Details
Entrez Gene ID: | 319454 |
Gene Name: | regulatory associated protein of MTOR, complex 1, opposite strand |
Gene Aliases: |
A430071A18Rik |
Location: |
Chr.11:119820811-119834365 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rptoros | NR_045312.1 | |||
| NR_045313.1 | ||||
| AK040159.1 | ||||
| AK086908.1 |

Genomic Map