Assay Details
Target Gene Details
Entrez Gene ID: | 445007 |
Gene Name: | nucleoporin 85 |
Gene Aliases: |
Pcnt1, frount |
Location: |
Chr.11:115564432-115583924 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup85 | NM_001002929.4 | NP_001002929.3 | ||
| AF498263.1 | AAM18529.1 | |||
| AK017632.1 | BAB30848.1 | |||
| AK187293.1 | ||||
| BC079856.1 | AAH79856.1 |
More Information
Set Membership: |
|

Genomic Map