Assay Details
Target Gene Details
Entrez Gene ID: | 67811 |
Gene Name: | polymerase (DNA-directed), delta interacting protein 2 |
Gene Aliases: |
1300003F06Rik, Pdip38 |
Location: |
Chr.11:78512296-78522736 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poldip2 | NM_026389.3 | NP_080665.1 | ||
| AK003702.1 | ||||
| AK165703.1 | ||||
| AK169503.1 | ||||
| AK211944.1 | ||||
| AK213405.1 | ||||
| BC006833.1 | AAH06833.1 | |||
| BC012879.1 | AAH12879.1 |

Genomic Map