Assay Details
Target Gene Details
Entrez Gene ID: | 170472 |
Gene Name: | RecQ protein-like 5 |
Gene Aliases: |
RecQ5, Recq5b, Recql5b |
Location: |
Chr.11:115892595-115933531 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Recql5 | NM_130454.2 | NP_569721.1 | ||
| AB038562.1 | BAD11129.1 | |||
| AB059999.1 | BAB79232.1 | |||
| AK029060.1 | ||||
| AK050965.1 | BAC34479.1 | |||
| AK083513.1 | ||||
| AK135177.1 | 1 | 189 |
More Information
Set Membership: |
|

Genomic Map