Assay Details
Target Gene Details
Entrez Gene ID: | 18103 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 2 |
Gene Aliases: |
NM23-H2, NM23B, nm23-M2 |
Location: |
Chr.11:93949814-93956256 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme2 | NM_008705.5 | 1 | 260 | NP_032731.1 |
| BC066995.1 | 1 | 260 | AAH66995.1 | |
| BY274368.1 |
More Information
Set Membership: |
|

Genomic Map