Assay Details
Target Gene Details
Entrez Gene ID: | 18453 |
Gene Name: | prolyl 4-hydroxylase, beta polypeptide |
Gene Aliases: |
ERp59, PDI, Pdia1, Thbp |
Location: |
Chr.11:120560304-120572936 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
More Information
Set Membership: |
|

Genomic Map