Assay Details
Target Gene Details
Entrez Gene ID: | 54125 |
Gene Name: | polymerase (DNA directed), mu |
Gene Aliases: |
AI462166, B230309I03Rik, Tdt-N |
Location: |
Chr.11:5827860-5838341 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polm | NM_017401.2 | NP_059097.2 | ||
| AF176098.1 | AAF27552.1 | |||
| AJ251804.2 | CAB71154.1 | |||
| BC056213.1 | AAH56213.1 |
More Information
Set Membership: |
|

Genomic Map