Assay Details
Target Gene Details
Entrez Gene ID: | 54125 |
Gene Name: | polymerase (DNA directed), mu |
Gene Aliases: |
AI462166, B230309I03Rik, Tdt-N |
Location: |
Chr.11:5827860-5838341 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polm | NM_017401.2 | 9 | 1470 | NP_059097.2 |
| AF176098.1 | 9 | 1243 | AAF27552.1 | |
| AJ251804.2 | 9 | 1214 | CAB71154.1 | |
| AK203359.1 | ||||
| BC056213.1 | 9 | 1262 | AAH56213.1 |
More Information
Set Membership: |
|

Genomic Map